Assay Details
Target Gene Details
Entrez Gene ID: | 104806 |
Gene Name: | Fanconi anemia, complementation group M |
Gene Aliases: |
AI427100, C730036B14Rik, D12Ertd364e |
Location: |
Chr.12:65074091-65132058 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fancm | NM_178912.3 | 18 | 4613 | NP_849243.2 |
| AK028941.1 | BAC26205.1 | |||
| AK173211.1 | 18 | 4633 | BAD32489.1 | |
| BC150785.1 | 18 | 4607 |
More Information
Set Membership: |
|

Genomic Map