Assay Details
Target Gene Details
Entrez Gene ID: | 73736 |
Gene Name: | FCF1 small subunit (SSU) processome component homolog (S. cerevisiae) |
Gene Aliases: |
1110008B24Rik |
Location: |
Chr.12:84970930-84983303 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcf1 | NM_028632.2 | NP_082908.2 | ||
| AK003561.1 | BAB22857.1 | |||
| BC033045.1 | AAH33045.1 |
More Information
Set Membership: |
|

Genomic Map