Assay Details
Target Gene Details
Entrez Gene ID: | 13411 |
Gene Name: | dynein, axonemal, heavy chain 11 |
Gene Aliases: |
Dnahc11, b2b1203Clo, b2b1279Clo, b2b1289Clo, b2b1727Clo, b2b598Clo, iv, lrd |
Location: |
Chr.12:117877982-118199043 on Build GRCm38 |
Assay Gene Location: | Within Exon 77 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnah11 | NM_010060.3 | 77 | 12680 | NP_034190.3 |
| AF183144.1 | 77 | 12680 | AAF07922.1 |
More Information
Set Membership: |
|

Genomic Map