Assay Details
Target Gene Details
Entrez Gene ID: | 110606 |
Gene Name: | farnesyltransferase, CAAX box, beta |
Gene Aliases: |
2010013E13Rik, AA409500 |
Location: |
Chr.12:76837398-76921412 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fntb | NM_145927.2 | 11 | 1161 | NP_666039.1 |
| AK089911.1 | 11 | 1190 | ||
| BC031417.1 | 11 | 1161 | AAH31417.1 |
More Information
Set Membership: |
|

Genomic Map