Assay Details
Target Gene Details
Entrez Gene ID: | 76044 |
Gene Name: | non-SMC condensin II complex, subunit G2 |
Gene Aliases: |
5830426I05Rik, Luzp5, Mtb, mCAP-G2 |
Location: |
Chr.12:116405357-116463532 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncapg2 | NM_133762.3 | 13 | 1506 | NP_598523.3 |
| AY455829.1 | 7 | 617 | AAR21588.1 | |
| BC008539.1 | 2 | 187 | AAH08539.1 | |
| BC076631.1 | 12 | 1316 | AAH76631.1 |

Genomic Map