Assay Details
Target Gene Details
Entrez Gene ID: | 72308 |
Gene Name: | BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae) |
Gene Aliases: |
2510002F24Rik, GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90 |
Location: |
Chr.12:112955631-113000707 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brf1 | NM_028193.3 | NP_082469.2 | ||
| BC037224.1 | AAH37224.1 | |||
| CF737042.1 |
More Information
Set Membership: |
|

Genomic Map