Assay Details
Target Gene Details
Entrez Gene ID: | 76044 |
Gene Name: | non-SMC condensin II complex, subunit G2 |
Gene Aliases: |
5830426I05Rik, Luzp5, Mtb, mCAP-G2 |
Location: |
Chr.12:116405357-116463532 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncapg2 | NM_133762.3 | 20 | 2519 | NP_598523.3 |
| AY455829.1 | 14 | 1630 | AAR21588.1 | |
| BC008539.1 | 9 | 1200 | AAH08539.1 | |
| BC076631.1 | 19 | 2329 | AAH76631.1 | |
| CK636577.1 | 4 | 340 |

Genomic Map