Assay Details
Target Gene Details
Entrez Gene ID: | 17977 |
Gene Name: | nuclear receptor coactivator 1 |
Gene Aliases: |
KAT13A, SRC-1, SRC1, bHLHe74 |
Location: |
Chr.12:4247362-4477210 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa1 | NM_010881.2 | 19 | 4522 | NP_035011.1 |
| AK035922.1 | 4 | 846 | BAC29244.1 | |
| BC068177.1 | 16 | 3830 | AAH68177.1 | |
| BC080866.1 | 19 | 4285 | AAH80866.1 | |
| U56920.1 | 18 | 4080 | AAB01228.1 | |
| U64606.1 | 16 | 3825 | AAB06177.1 | |
| U64828.1 | 19 | 4435 | AAB38841.1 |
More Information
Set Membership: |
|

Genomic Map