Assay Details
Target Gene Details
Entrez Gene ID: | 67732 |
Gene Name: | isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae) |
Gene Aliases: |
1500019E20Rik, 4833421E05Rik |
Location: |
Chr.12:21316360-21323607 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iah1 | NM_026347.3 | 1 | 59 | NP_080623.2 |
| AK005287.1 | 1 | 60 | BAB23934.1 | |
| AK011299.1 | 1 | 60 | ||
| AK014743.1 | 1 | 37 | ||
| BC060949.1 | 1 | 62 | AAH60949.1 | |
| BC087901.1 | AAH87901.1 |
More Information
Set Membership: |
|

Genomic Map