Assay Details
Target Gene Details
Entrez Gene ID: | 17977 |
Gene Name: | nuclear receptor coactivator 1 |
Gene Aliases: |
KAT13A, SRC-1, SRC1, bHLHe74 |
Location: |
Chr.12:4247362-4477210 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa1 | NM_010881.2 | NP_035011.1 | ||
| BC068177.1 | AAH68177.1 | |||
| BC080866.1 | AAH80866.1 | |||
| U56920.1 | AAB01228.1 | |||
| U64606.1 | AAB06177.1 | |||
| U64828.1 | AAB38841.1 |
More Information
Set Membership: |
|

Genomic Map