Assay Details
Target Gene Details
Entrez Gene ID: | 76826 |
Gene Name: | nucleotide binding protein-like |
Gene Aliases: |
2410170E07Rik |
Location: |
Chr.12:52097746-52310970 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nubpl | NM_029760.2 | NP_084036.2 | ||
| AI118524.1 | ||||
| AK010827.1 | BAB27209.1 | |||
| AK141257.1 | BAE24620.1 | |||
| AK152377.1 | BAE31166.1 | |||
| BC062175.1 | AAH62175.1 |
More Information
Set Membership: |
|

Genomic Map