Assay Details
Target Gene Details
Entrez Gene ID: | 56217 |
Gene Name: | membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5) |
Gene Aliases: |
3830420B02Rik, AI255216, AI644496, Pals1 |
Location: |
Chr.12:78748912-78840877 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp5 | NM_019579.3 | NP_062525.1 | ||
| AF199008.1 | AAF63789.1 | |||
| AK014444.1 | ||||
| AK032731.1 | ||||
| AK133187.1 | ||||
| AK162014.1 | ||||
| BB858385.1 | ||||
| BC138624.1 |
More Information
Set Membership: |
|

Genomic Map