Assay Details
Target Gene Details
Entrez Gene ID: | 13435 |
Gene Name: | DNA methyltransferase 3A |
Gene Aliases: |
MmuIIIA |
Location: |
Chr.12:3804535-3914443 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnmt3a | NM_001271753.1 | NP_001258682.1 | ||
| NM_007872.4 | NP_031898.1 | |||
| NM_153743.4 | NP_714965.1 | |||
| AF068625.2 | AAC40177.2 | |||
| AF480164.1 | AAN40038.1 | |||
| AK077792.1 | BAC37013.1 | |||
| AK090132.1 | BAC41110.1 | |||
| AK147263.1 | BAE27806.1 | |||
| AK147627.1 | BAE28033.1 | |||
| AK147642.1 | BAE28043.1 | |||
| AK147676.1 | BAE28067.1 | |||
| AK157792.1 | BAE34200.1 | |||
| BC007466.1 | AAH07466.1 |
More Information
Set Membership: |
|

Genomic Map