Assay Details
Target Gene Details
Entrez Gene ID: | 217715 |
Gene Name: | eukaryotic translation initiation factor 2B, subunit 2 beta |
Gene Aliases: |
AA409345, C85417, EIF-2Bbeta, EIF2B |
Location: |
Chr.12:85219475-85226628 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2b2 | NM_145445.3 | NP_663420.1 | ||
| AK049731.1 | BAC33897.1 | |||
| AK076195.1 | BAC36248.1 | |||
| AK194280.1 | ||||
| BC003326.1 | AAH03326.1 |
More Information
Set Membership: |
|

Genomic Map