Assay Details
Target Gene Details
Entrez Gene ID: | 16593 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
AI874768, Kns2 |
Location: |
Chr.12:111758849-111807844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klc1 | NM_001025360.2 | NP_001020531.2 | ||
| NM_001025361.2 | NP_001020532.2 | |||
| NM_001025362.2 | NP_001020533.2 | |||
| NM_001081959.1 | NP_001075428.1 | |||
| AK014256.1 | ||||
| AK031309.1 | BAC27341.1 | |||
| AV319299.2 | ||||
| BB835417.1 | ||||
| BC055744.1 | AAH55744.1 |
Target Gene Details
Entrez Gene ID: | 74335 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 3 |
Gene Aliases: |
4432412E01Rik, AI182522, AW537713 |
Location: |
Chr.12:111803192-111813841 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc3 | NM_028875.2 | NP_083151.1 | ||
| AK014491.1 | BAB29391.1 | |||
| BC043073.1 | AAH43073.1 |
More Information
Set Membership: |
|

Genomic Map