Assay Details
Target Gene Details
Entrez Gene ID: | 54451 |
Gene Name: | cleavage and polyadenylation specificity factor 3 |
Gene Aliases: |
- |
Location: |
Chr.12:21286297-21315056 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf3 | NM_018813.3 | NP_061283.2 | ||
| AF203969.1 | AAF19420.1 | |||
| AK149953.1 | BAE29190.1 | |||
| AK157205.1 | BAE33998.1 | |||
| AK158036.1 | ||||
| AK170842.1 | BAE42066.1 | |||
| AK171838.1 | ||||
| BC023297.1 | AAH23297.1 | |||
| BC100569.1 | AAI00570.1 | |||
| CF725936.1 |
More Information
Set Membership: |
|

Genomic Map