Assay Details
Target Gene Details
Entrez Gene ID: | 54451 |
Gene Name: | cleavage and polyadenylation specificity factor 3 |
Gene Aliases: |
- |
Location: |
Chr.12:21286297-21315056 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf3 | NM_018813.3 | 5 | 585 | NP_061283.2 |
| AF203969.1 | 5 | 438 | AAF19420.1 | |
| AK149953.1 | 5 | 512 | BAE29190.1 | |
| AK157205.1 | 5 | 527 | BAE33998.1 | |
| AK158036.1 | 5 | 497 | ||
| AK170842.1 | 5 | 470 | BAE42066.1 | |
| AK171838.1 | 5 | 516 | ||
| BC023297.1 | 5 | 471 | AAH23297.1 | |
| BC100569.1 | 5 | 483 | AAI00570.1 | |
| CF725936.1 | 3 | 194 |
More Information
Set Membership: |
|

Genomic Map