Assay Details
Target Gene Details
Entrez Gene ID: | 54451 |
Gene Name: | cleavage and polyadenylation specificity factor 3 |
Gene Aliases: |
- |
Location: |
Chr.12:21286297-21315056 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf3 | NM_018813.3 | 8 | 1042 | NP_061283.2 |
| AF203969.1 | 8 | 895 | AAF19420.1 | |
| AK010566.1 | 1 | 109 | BAB27031.1 | |
| AK149953.1 | 8 | 969 | BAE29190.1 | |
| AK157205.1 | 8 | 984 | BAE33998.1 | |
| AK158036.1 | 8 | 954 | ||
| AK170842.1 | 8 | 927 | BAE42066.1 | |
| AK171838.1 | 8 | 973 | ||
| AK197891.1 | ||||
| AK212230.1 | ||||
| AK212645.1 | ||||
| BC023297.1 | 8 | 928 | AAH23297.1 | |
| BC100569.1 | 8 | 940 | AAI00570.1 | |
| CF725936.1 | 6 | 651 |
More Information
Set Membership: |
|

Genomic Map