Assay Details
Target Gene Details
Entrez Gene ID: | 217480 |
Gene Name: | diacylglycerol kinase, beta |
Gene Aliases: |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
Location: |
Chr.12:37880168-38634239 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkb | NM_178681.4 | 11 | 1382 | NP_848796.2 |
| AK032509.1 | 11 | 1541 | ||
| AK038544.1 | 11 | 1110 | BAC30034.1 | |
| AK044516.1 | 11 | 1192 | BAC31960.1 | |
| AK047682.1 | 12 | 1318 | BAC33123.2 | |
| AK083222.1 | 12 | 1393 | BAC38814.1 | |
| AK122355.1 | 10 | 870 | BAC65637.1 | |
| AK139408.1 | 11 | 1383 | BAE23997.1 | |
| BC070461.1 | 11 | 1214 | AAH70461.1 |
More Information
Set Membership: |
|

Genomic Map