Assay Details
Target Gene Details
Entrez Gene ID: | 217480 |
Gene Name: | diacylglycerol kinase, beta |
Gene Aliases: |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
Location: |
Chr.12:37880168-38634239 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkb | NM_178681.4 | 14 | 1626 | NP_848796.2 |
| AK032509.1 | 14 | 1785 | ||
| AK038544.1 | 14 | 1354 | BAC30034.1 | |
| AK044516.1 | BAC31960.1 | |||
| AK047682.1 | BAC33123.2 | |||
| AK083222.1 | 15 | 1637 | BAC38814.1 | |
| AK122355.1 | 13 | 1114 | BAC65637.1 | |
| AK139408.1 | 14 | 1627 | BAE23997.1 | |
| BC070461.1 | 14 | 1458 | AAH70461.1 |
More Information
Set Membership: |
|

Genomic Map