Assay Details
Target Gene Details
Entrez Gene ID: | 217480 |
Gene Name: | diacylglycerol kinase, beta |
Gene Aliases: |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
Location: |
Chr.12:37880168-38634239 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkb | NM_178681.4 | 18 | 1972 | NP_848796.2 |
| AK032509.1 | 18 | 2131 | ||
| AK122355.1 | 17 | 1460 | BAC65637.1 | |
| AK139408.1 | 18 | 1973 | BAE23997.1 | |
| BC070461.1 | 18 | 1804 | AAH70461.1 |
More Information
Set Membership: |
|

Genomic Map