Assay Details
Target Gene Details
Entrez Gene ID: | 17977 |
Gene Name: | nuclear receptor coactivator 1 |
Gene Aliases: |
KAT13A, SRC-1, SRC1, bHLHe74 |
Location: |
Chr.12:4247362-4477210 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa1 | NM_010881.2 | NP_035011.1 | ||
| CJ066829.1 | ||||
| U64828.1 | AAB38841.1 |
More Information
Set Membership: |
|

Genomic Map