Assay Details
Target Gene Details
Entrez Gene ID: | 16593 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
AI874768, Kns2 |
Location: |
Chr.12:111758849-111807844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klc1 | NM_001025358.2 | NP_001020529.2 | ||
| NM_001025359.2 | NP_001020530.2 | |||
| NM_001025360.2 | NP_001020531.2 | |||
| NM_001025361.2 | NP_001020532.2 | |||
| NM_001025362.2 | NP_001020533.2 | |||
| NM_001025363.2 | NP_001020534.2 | |||
| NM_001081959.1 | NP_001075428.1 | |||
| NM_008450.2 | NP_032476.2 | |||
| AF055665.1 | AAC27740.1 | |||
| AK014256.1 | ||||
| AK031309.1 | BAC27341.1 | |||
| AY753300.1 | AAV30550.1 | |||
| BC055744.1 | AAH55744.1 | |||
| BG094009.1 | ||||
| BU922789.1 | ||||
| CK032061.1 |
More Information
Set Membership: |
|

Genomic Map