Assay Details
Target Gene Details
Entrez Gene ID: | 73834 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit D |
Gene Aliases: |
1110004P10Rik, Atp6m, VATD, Vma8 |
Location: |
Chr.12:78842982-78861638 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1d | NM_023721.2 | NP_076210.1 | ||
| AF298810.1 | AAG30225.1 | |||
| AK003446.1 | BAB22795.1 | |||
| AK146129.1 | BAE26921.1 | |||
| AK146448.1 | BAE27180.1 | |||
| AK150981.1 | BAE30008.1 | |||
| AK152367.1 | BAE31157.1 | |||
| AK153250.1 | BAE31841.1 | |||
| AK165761.1 | BAE38369.1 | |||
| BC033457.1 | AAH33457.1 |
More Information
Set Membership: |
|

Genomic Map