Assay Details
Target Gene Details
Entrez Gene ID: | 217480 |
Gene Name: | diacylglycerol kinase, beta |
Gene Aliases: |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
Location: |
Chr.12:37880168-38634239 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkb | NM_178681.4 | 22 | 2481 | NP_848796.2 |
| AK032509.1 | 22 | 2640 | ||
| AK047785.1 | 3 | 302 | BAC33156.1 | |
| AK122355.1 | 21 | 1969 | BAC65637.1 | |
| AK139408.1 | 22 | 2482 | BAE23997.1 | |
| BC070461.1 | 22 | 2313 | AAH70461.1 |
More Information
Set Membership: |
|

Genomic Map