Assay Details
Target Gene Details
Entrez Gene ID: | 271005 |
Gene Name: | kelch domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.12:69226917-69284646 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klhdc1 | NM_178253.5 | NP_839984.1 | ||
AK144621.1 | BAE25974.1 | |||
AY238938.1 | AAO88963.1 | |||
BC119567.2 | ||||
BC119568.2 |
