Assay Details
Target Gene Details
Entrez Gene ID: | 104836 |
Gene Name: | Casitas B-lineage lymphoma-like 1 |
Gene Aliases: |
AI467391, Hakai, c-Cbl-like |
Location: |
Chr.12:31484829-31500741 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbll1 | NM_001253847.1 | NP_001240776.1 | ||
| NM_001253848.1 | NP_001240777.1 | |||
| NM_134048.2 | NP_598809.1 | |||
| AF167441.1 | AAF89617.1 | |||
| AK049141.1 | BAC33568.1 | |||
| AK051737.1 | ||||
| AK166017.1 | BAE38522.1 | |||
| BB651673.1 | ||||
| BC019529.1 | AAH19529.1 |
More Information
Set Membership: |
|

Genomic Map