Assay Details
Target Gene Details
Entrez Gene ID: | 16413 |
Gene Name: | integrin beta 1 binding protein 1 |
Gene Aliases: |
AI449260, AU019480, Icap1 |
Location: |
Chr.12:21267246-21286279 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb1bp1 | NM_008403.4 | NP_032429.1 | ||
| AJ001373.1 | CAA04706.1 | |||
| AK002786.1 | BAB22357.1 | |||
| AK004172.1 | BAB23206.1 | |||
| AK007635.1 | BAB25151.1 | |||
| AK075577.1 | BAC35832.1 | |||
| AK089937.1 | BAC41007.1 | |||
| AK147929.1 | BAE28236.1 | |||
| BC028772.1 | AAH28772.1 |
More Information
Set Membership: |
|

Genomic Map