Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | NP_001074922.1 | ||
| NM_001286079.2 | NP_001273008.1 | |||
| NM_001286080.2 | NP_001273009.1 | |||
| NM_008697.4 | NP_032723.2 | |||
| NR_104397.2 | ||||
| NR_104398.2 | ||||
| AK129394.1 | BAC98204.1 | |||
| AK134860.1 | ||||
| AK147187.1 | ||||
| AK162150.1 | ||||
| AK167264.1 | ||||
| AY515727.1 | AAS87211.1 | |||
| BC137789.1 | ||||
| BC144913.1 | ||||
| U40342.1 | AAA83234.1 |

Genomic Map