Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | 23 | 5358 | NP_001074922.1 |
| NM_001286079.2 | 24 | 5540 | NP_001273008.1 | |
| NM_001286080.2 | 25 | 5587 | NP_001273009.1 | |
| NM_008697.4 | 25 | 5587 | NP_032723.2 | |
| NR_104397.2 | 25 | 5587 | ||
| NR_104398.2 | 23 | 3431 | ||
| AK129394.1 | 25 | 5477 | BAC98204.1 | |
| AK134860.1 | 23 | 3431 | ||
| AK147187.1 | 8 | 2343 | ||
| AK167264.1 | 21 | 4453 | ||
| AY515727.1 | 23 | 5337 | AAS87211.1 | |
| BC137789.1 | 24 | 5425 | ||
| BC144913.1 | 25 | 5472 | ||
| U40342.1 | 25 | 5494 | AAA83234.1 |

Genomic Map