Assay Details
Target Gene Details
Entrez Gene ID: | 17933 |
Gene Name: | myelin transcription factor 1-like |
Gene Aliases: |
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106 |
Location: |
Chr.12:29528311-29923216 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myt1l | NM_001093775.1 | 14 | 2789 | NP_001087244.1 |
| NM_001093776.1 | 14 | 2695 | NP_001087245.1 | |
| NM_001093778.1 | 14 | 2780 | NP_001087247.1 | |
| NM_008666.3 | 14 | 2783 | NP_032692.2 | |
| AB093283.1 | 11 | 2142 | BAC41467.1 | |
| AB212898.1 | 13 | 2387 | BAE48255.1 | |
| AF004295.1 | 14 | 2602 | AAC53457.1 | |
| AK049967.1 | 5 | 624 | BAC34010.1 | |
| AK076608.1 | 2 | 220 | BAC36413.1 | |
| BC094438.1 | 10 | 2020 | ||
| BC131677.1 | 13 | 2384 | ||
| U86338.1 | 14 | 2511 | AAC53157.1 |
More Information
Set Membership: |
|

Genomic Map