Assay Details
Target Gene Details
Entrez Gene ID: | 217837 |
Gene Name: | inositol 1,3,4-triphosphate 5/6 kinase |
Gene Aliases: |
BC031182 |
Location: |
Chr.12:102568583-102704869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpk1 | NM_172584.3 | NP_766172.1 | ||
| AK038913.1 | BAC30166.1 | |||
| BC056464.1 | AAH56464.1 |
More Information
Set Membership: |
|

Genomic Map