Assay Details
Target Gene Details
Entrez Gene ID: | 74724 |
Gene Name: | RIKEN cDNA 4930512B01 gene |
Gene Aliases: |
4930472D12Rik |
Location: |
Chr.12:69790344-69841880 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930512B01Rik | NR_033573.1 | |||
| AK015558.1 |
Target Gene Details
Entrez Gene ID: | 399510 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 5 |
Gene Aliases: |
4432415E19Rik, GCKR, KHS, MAPKKKK5 |
Location: |
Chr.12:69803757-69893163 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map4k5 | NM_201519.2 | NP_958927.2 | ||
| AK019468.1 | BAB31739.1 | |||
| AK032123.1 | ||||
| AK053775.1 | BAC35517.1 | |||
| AK084891.1 | BAC39305.1 | |||
| BC040381.1 | AAH40381.2 | |||
| BC048173.1 |

Genomic Map