Assay Details
Target Gene Details
Entrez Gene ID: | 17933 |
Gene Name: | myelin transcription factor 1-like |
Gene Aliases: |
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106 |
Location: |
Chr.12:29528311-29923216 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myt1l | NM_001093775.1 | NP_001087244.1 | ||
| NM_001093776.1 | NP_001087245.1 | |||
| NM_001093778.1 | NP_001087247.1 | |||
| NM_008666.3 | NP_032692.2 | |||
| AB212898.1 | BAE48255.1 | |||
| AF004295.1 | AAC53457.1 | |||
| AK012660.1 | ||||
| AK036205.1 | ||||
| AK049624.1 | ||||
| BC131677.1 | ||||
| BE653129.1 | ||||
| CB521730.1 | ||||
| CD352642.1 | ||||
| U86338.1 | AAC53157.1 |
More Information
Set Membership: |
|

Genomic Map