Assay Details
Target Gene Details
Entrez Gene ID: | 69706 |
Gene Name: | leucine rich repeat protein 1 |
Gene Aliases: |
2410005L11Rik, AU021780, LRR-1, Ppil5 |
Location: |
Chr.12:69168814-69179010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrr1 | NM_001081406.1 | NP_001074875.1 | ||
| AK010419.1 | BAB26922.1 | |||
| AK144741.1 | ||||
| BC022648.1 | AAH22648.1 | |||
| BC117053.1 | AAI17054.1 | |||
| BC117055.1 | AAI17056.1 |
More Information
Set Membership: |
|

Genomic Map