Assay Details
Target Gene Details
Entrez Gene ID: | 72371 |
Gene Name: | RIKEN cDNA 2210408I21 gene |
Gene Aliases: |
- |
Location: |
Chr.13:77135536-77613679 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2210408I21Rik | NM_001145676.1 | 5 | 1151 | NP_001139148.1 |
| AK143619.1 | 5 | 1089 | BAE25469.1 | |
| AK165312.1 | 5 | 1148 | BAE38131.1 | |
| BC151066.1 | 5 | 1133 | ||
| BC157949.1 | 5 | 1133 |

Genomic Map