Assay Details
Target Gene Details
Entrez Gene ID: | 319565 |
Gene Name: | spectrin repeat containing, nuclear envelope 2 |
Gene Aliases: |
6820443O06Rik, AW546258, D12Ertd777e, NUA, Nesp2g, mKIAA1011 |
Location: |
Chr.12:75818318-76110928 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syne2 | NM_001005510.2 | 24 | 3285 | NP_001005510.2 |

Genomic Map