Assay Details
Target Gene Details
Entrez Gene ID: | 217449 |
Gene Name: | trafficking protein particle complex 12 |
Gene Aliases: |
CGI-87, D930014A20Rik, Ttc15 |
Location: |
Chr.12:28690620-28750488 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc12 | NM_001161410.1 | NP_001154882.1 | ||
| NM_178811.4 | NP_848926.3 | |||
| AK049172.1 | BAC33585.1 | |||
| AK132166.1 | ||||
| AK147985.1 | BAE28268.1 | |||
| BC002235.1 | AAH02235.1 | |||
| BC024077.1 | AAH24077.1 | |||
| BC030887.1 | AAH30887.1 |
More Information
Set Membership: |
|

Genomic Map