Assay Details
Target Gene Details
Entrez Gene ID: | 53612 |
Gene Name: | vesicle transport through interaction with t-SNAREs 1B |
Gene Aliases: |
AU015348, GES30, MVti1b, SNARE, Vti1-rp1 |
Location: |
Chr.12:79156017-79172688 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vti1b | NM_016800.3 | 4 | 581 | NP_058080.2 |
| AF035208.1 | 4 | 468 | AAC23483.1 | |
| AF035821.1 | 4 | 454 | AAD02039.1 | |
| AK002289.1 | 4 | 573 | ||
| AK160624.1 | 3 | 515 | BAE35923.1 | |
| BC009810.1 | 4 | 511 | AAH09810.1 | |
| BC080204.1 | 4 | 557 | AAH80204.1 | |
| BQ177030.1 | ||||
| CX239415.1 | 4 | 558 |
More Information
Set Membership: |
|

Genomic Map