Assay Details
Target Gene Details
Entrez Gene ID: | 238384 |
Gene Name: | solute carrier family 24 (sodium/potassium/calcium exchanger), member 4 |
Gene Aliases: |
A930002M03Rik, AW492432, NCKX4 |
Location: |
Chr.12:102128652-102269083 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc24a4 | NM_172152.2 | NP_742164.1 | ||
| AK044239.1 | BAC31835.1 | |||
| AK044368.1 | BAC31887.1 | |||
| AY156046.1 | AAN37415.1 | |||
| BC137945.1 | ||||
| BC171939.1 |
More Information
Set Membership: |
|

Genomic Map