Assay Details
Target Gene Details
Entrez Gene ID: | 104718 |
Gene Name: | tetratricopeptide repeat domain 7B |
Gene Aliases: |
AA408451, Ttc7l1 |
Location: |
Chr.12:100300770-100520822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc7b | NM_001033213.1 | NP_001028385.1 | ||
| AK046740.1 |

Genomic Map