Assay Details
Target Gene Details
Entrez Gene ID: | 66975 |
Gene Name: | trafficking protein particle complex 13 |
Gene Aliases: |
2410002O22Rik, 2610524F24Rik, AI451892, AU017229 |
Location: |
Chr.13:104142153-104178703 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc13 | NM_001093759.1 | NP_001087228.1 | ||
| NM_001093760.1 | NP_001087229.1 | |||
| NM_025879.2 | NP_080155.3 | |||
| AK010346.1 | BAB26869.2 | |||
| AK013956.1 | BAB29083.2 | |||
| AK084373.1 | BAC39168.1 | |||
| AK149832.1 | BAE29111.1 | |||
| AK163342.1 | BAE37309.1 | |||
| AK167749.1 | BAE39785.1 | |||
| BC021756.1 |

Genomic Map