Assay Details
Target Gene Details
Entrez Gene ID: | 17252 |
Gene Name: | retinol dehydrogenase 11 |
Gene Aliases: |
2610319N22Rik, AI428145, AU045252, Arsdr1, C85936, CGI-82, HCBP12, M42C60, Mdt1, Psdr1, SCALD, UBE-1c1, Ube-1c |
Location: |
Chr.12:79174335-79191819 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh11 | NM_021557.5 | NP_067532.2 | ||
| AB030503.1 | BAA82656.1 | |||
| AB030504.1 | BAA82657.1 | |||
| AB035959.1 | BAA88521.1 | |||
| AF474027.1 | AAL79910.1 | |||
| AK004413.1 | BAB23296.1 | |||
| AK012056.1 | ||||
| AK135443.1 | BAE22534.1 | |||
| AY039032.1 | AAK91516.1 | |||
| BC018261.1 | AAH18261.1 |
More Information
Set Membership: |
|

Genomic Map