Assay Details
Target Gene Details
Entrez Gene ID: | 217593 |
Gene Name: | solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21 |
Gene Aliases: |
9930033G19Rik, A630030I10, BB158148 |
Location: |
Chr.12:56712634-57197472 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a21 | NM_001167976.1 | NP_001161448.1 | ||
| NM_172577.3 | NP_766165.2 | |||
| AK036988.1 | BAC29659.1 | |||
| AK041694.1 | BAC31036.1 | |||
| AK045180.1 | ||||
| AK162379.1 | BAE36882.1 | |||
| AK167535.1 | ||||
| BC057980.1 | AAH57980.1 | |||
| EU180236.1 |
More Information
Set Membership: |
|

Genomic Map