Assay Details
Target Gene Details
Entrez Gene ID: | 217593 |
Gene Name: | solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21 |
Gene Aliases: |
9930033G19Rik, A630030I10, BB158148 |
Location: |
Chr.12:56712634-57197472 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a21 | NM_001167976.1 | 9 | 827 | NP_001161448.1 |
| NM_172577.3 | 8 | 1270 | NP_766165.2 | |
| AK036988.1 | 8 | 1010 | BAC29659.1 | |
| AK041694.1 | 8 | 1271 | BAC31036.1 | |
| AK045180.1 | 8 | 1096 | ||
| AK162379.1 | 8 | 1097 | BAE36882.1 | |
| AK167535.1 | 8 | 978 | ||
| BC057980.1 | 8 | 1094 | AAH57980.1 | |
| EU180236.1 | 9 | 827 |
More Information
Set Membership: |
|

Genomic Map