Assay Details
Target Gene Details
Entrez Gene ID: | 320808 |
Gene Name: | DDB1 and CUL4 associated factor 5 |
Gene Aliases: |
9430020B07Rik, AI325023, AI430035, AI848110, AW124614, BCRG2, BCRP2, Wdr22 |
Location: |
Chr.12:80335847-80436601 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf5 | NM_177267.4 | NP_796241.3 | ||
| AK122561.1 | BAC65843.1 | |||
| AK137579.1 | BAE23416.1 | |||
| AK165096.1 | ||||
| BC042567.1 | AAH42567.1 | |||
| BC046813.1 | AAH46813.2 |
More Information
Set Membership: |
|

Genomic Map