Assay Details
Target Gene Details
Entrez Gene ID: | 77974 |
Gene Name: | retinol dehydrogenase 12 |
Gene Aliases: |
A930033N07Rik |
Location: |
Chr.12:79208914-79222661 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh12 | NM_030017.3 | NP_084293.1 | ||
| AK020927.1 | BAB32258.1 | |||
| AK039233.1 | BAC30288.1 | |||
| BC016204.1 | AAH16204.1 |
More Information
Set Membership: |
|

Genomic Map