Assay Details
Target Gene Details
Entrez Gene ID: | 319565 |
Gene Name: | spectrin repeat containing, nuclear envelope 2 |
Gene Aliases: |
6820443O06Rik, AW546258, D12Ertd777e, NUA, Nesp2g, mKIAA1011 |
Location: |
Chr.12:75818318-76110928 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syne2 | NM_001005510.2 | 7 | 708 | NP_001005510.2 |
| AF435012.1 | 7 | 692 | AAL33549.1 | |
| AK052521.1 | 7 | 677 | BAC35023.1 | |
| AK078542.1 | 7 | 709 | ||
| AK084394.1 | 8 | 888 | BAC39173.1 | |
| AK084811.1 | 7 | 708 | ||
| AK145252.1 | 7 | 709 |

Genomic Map