Assay Details
Target Gene Details
Entrez Gene ID: | 20663 |
Gene Name: | son of sevenless homolog 2 (Drosophila) |
Gene Aliases: |
SOS-2, mSOS-2 |
Location: |
Chr.12:69583761-69681852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sos2 | NM_001135559.1 | NP_001129031.1 | ||
| Z11664.1 | CAA77732.1 |
More Information
Set Membership: |
|

Genomic Map