Assay Details
Target Gene Details
Entrez Gene ID: | 627191 |
Gene Name: | synapse differentiation inducing 1 like |
Gene Aliases: |
DSPC1, Gm261, Tmem90a |
Location: |
Chr.12:84677277-84698907 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syndig1l | NM_001033334.2 | NP_001028506.1 | ||
| AK140195.1 | BAE24274.1 | |||
| AY993933.1 | AAY42815.1 | |||
| AY993934.1 | AAY42816.1 | |||
| BC132221.1 | ||||
| BC138501.1 |

Genomic Map